Learn More
Agilent Technologies Agilent Technologies Quick Connect Capillary, Stainless Steel, 0.12 mm ID, 150 mm
Catalog No. NC3981643
Encompass
This stainless steel Quick Connect capillary, with a 0.12 mm inner diameter and 150 mm length, is engineered for high-speed analytical applications in LC instruments. Its innovative, finger-tight, high-pressure design ensures zero-dead-volume connections, preventing issues like peak broadening and loss of resolution. It is stable up to 1300 bar and is designed for use with specific Quick Connect fittings.
  • Designed for analytical compatibility.
  • Quick Connect capillary subtype.
  • Features Quick Connect fitting type.
  • Left end has no fitting.
  • Right end includes SX fitting (Swagelok, 1.6 mm ID, stainless steel, extra long screw).
  • Right end fitting is non-swaged.
  • Optimized for liquid chromatography (LC) technique.
  • Ensures zero-dead-volume connections for optimal chromatographic performance.
Catalog No. NC3981643 Supplier Agilent Technologies Supplier No. 55001172
This stainless steel Quick Connect capillary, with a 0.12 mm inner diameter and 150 mm length, is engineered for high-speed analytical applications in LC instruments. Its innovative, finger-tight, high-pressure design ensures zero-dead-volume connections, preventing issues like peak broadening and loss of resolution. It is stable up to 1300 bar and is designed for use with specific Quick Connect fittings.
  • Designed for analytical compatibility.
  • Quick Connect capillary subtype.
  • Features Quick Connect fitting type.
  • Left end has no fitting.
  • Right end includes SX fitting (Swagelok, 1.6 mm ID, stainless steel, extra long screw).
  • Right end fitting is non-swaged.
  • Optimized for liquid chromatography (LC) technique.
  • Ensures zero-dead-volume connections for optimal chromatographic performance.