Learn More
Agilent Technologies Agilent Technologies Stay Safe Cap, S60, 4 Ports, for Spectroscopy
Catalog No. NC3981675
Encompass
This Stay Safe cap for spectroscopy prevents harmful acid vapors from escaping waste bottles. It features an S60 thread, 4 ports, 4 x barb fitting 1/4–28 male to 1/8 in, and all necessary screw plugs. The cap protects lab personnel from toxic chemicals and maintains solvent consistency.
  • Designed for bottles/vessels with S60 thread.
  • Minimizes harmful vapors, restricting solvents for a safer work environment.
  • Maintains solvent consistency, preventing mobile phase changes and reducing waste.
  • Stops dust and air contaminants for reliable chromatography results.
  • Includes a venting valve with a six–month time strip for replacement.
  • Enables vapor–free tubing connections; PFA fittings and PTFE screw plugs.
  • Reduces methanol evaporation by up to 85% compared to standard caps.
Catalog No. NC3981675 Supplier Agilent Technologies Supplier No. 50431326
This Stay Safe cap for spectroscopy prevents harmful acid vapors from escaping waste bottles. It features an S60 thread, 4 ports, 4 x barb fitting 1/4–28 male to 1/8 in, and all necessary screw plugs. The cap protects lab personnel from toxic chemicals and maintains solvent consistency.
  • Designed for bottles/vessels with S60 thread.
  • Minimizes harmful vapors, restricting solvents for a safer work environment.
  • Maintains solvent consistency, preventing mobile phase changes and reducing waste.
  • Stops dust and air contaminants for reliable chromatography results.
  • Includes a venting valve with a six–month time strip for replacement.
  • Enables vapor–free tubing connections; PFA fittings and PTFE screw plugs.
  • Reduces methanol evaporation by up to 85% compared to standard caps.