Learn More
Agilent Technologies Agilent Technologies Vial, 2 mL, Amber, Screw Top, Write-on Spot
Catalog No. NC3981668
Encompass
These 2 mL amber screw top vials with a write-on spot are designed for GC LC techniques. Manufactured from analytical grade glass (types 33 and 51), they ensure samples are not compromised by metal content or analyte removal. These vials offer fast cap manipulation and are tested for compatibility with over 10 instrument brands, preventing sample loss and autosampler damage.
  • Reduced labor with caps screwing/unscrewing up to 30% faster.
  • Tested for compatibility with over 10 instrument brands.
  • Consistent lot-to-lot performance and rigorous quality control.
  • Prevents sample loss, contamination, and damage to autosampler needles.
  • Constructed from analytical grades 33 and 51 glass.
  • Suitable for LC and LC/MS applications.
  • Certified for quality.
  • Features amber glass and a write-on spot for labeling.
  • Vial dimensions: 32 mm × 12 mm.
Catalog No. NC3981668 Supplier Agilent Technologies Supplier No. 51906115
These 2 mL amber screw top vials with a write-on spot are designed for GC LC techniques. Manufactured from analytical grade glass (types 33 and 51), they ensure samples are not compromised by metal content or analyte removal. These vials offer fast cap manipulation and are tested for compatibility with over 10 instrument brands, preventing sample loss and autosampler damage.
  • Reduced labor with caps screwing/unscrewing up to 30% faster.
  • Tested for compatibility with over 10 instrument brands.
  • Consistent lot-to-lot performance and rigorous quality control.
  • Prevents sample loss, contamination, and damage to autosampler needles.
  • Constructed from analytical grades 33 and 51 glass.
  • Suitable for LC and LC/MS applications.
  • Certified for quality.
  • Features amber glass and a write-on spot for labeling.
  • Vial dimensions: 32 mm × 12 mm.