Learn More
Agilent Technologies Agilent Technologies Vial Convenience Pack, 2 mL, Amber, PTFE/Silicone/PTFE Septa
Catalog No. NC3981638
Encompass
Includes 500 amber screw top vials with write-on spots, green polypropylene caps, and PTFE/red silicone septa. High-quality amber glass provides excellent visibility and chemical resistance. Tri-layer PTFE/silicone/PTFE septa ensure superior sealing and chemical inertness, even with dramatic temperature swings. Ideal for GC & LC applications, it supports drug discovery, stability testing, & quality control.
  • 500 amber vials with write-on spots & green caps.
  • High-quality amber glass.
  • Tri-layer PTFE/silicone/PTFE septa.
  • Septa for dramatic temperature swings & enhanced re-sealability.
  • Suitable for GC & LC applications.
  • Ideal for drug discovery, stability testing, & quality control.
Catalog No. NC3981638 Supplier Agilent Technologies Supplier No. 51820737
Includes 500 amber screw top vials with write-on spots, green polypropylene caps, and PTFE/red silicone septa. High-quality amber glass provides excellent visibility and chemical resistance. Tri-layer PTFE/silicone/PTFE septa ensure superior sealing and chemical inertness, even with dramatic temperature swings. Ideal for GC & LC applications, it supports drug discovery, stability testing, & quality control.
  • 500 amber vials with write-on spots & green caps.
  • High-quality amber glass.
  • Tri-layer PTFE/silicone/PTFE septa.
  • Septa for dramatic temperature swings & enhanced re-sealability.
  • Suitable for GC & LC applications.
  • Ideal for drug discovery, stability testing, & quality control.