Learn More
Agilent Technologies Agilent Technologies Vial, Crimp Top, Headspace, 10 mL, Flat Bottom
Catalog No. NC3981645
Encompass
This vial is a crimp top headspace vial with a 10 mL capacity and a flat bottom, commonly used in laboratory settings for various applications.
  • Features a crimp top design.
  • Suitable for headspace applications.
  • Has a 10 mL capacity.
  • Constructed with clear material.
  • Designed with a flat bottom.
  • Supplied in quantities of 1000 per case.
Catalog No. NC3981645 Supplier Agilent Technologies Supplier No. 80100179
This vial is a crimp top headspace vial with a 10 mL capacity and a flat bottom, commonly used in laboratory settings for various applications.
  • Features a crimp top design.
  • Suitable for headspace applications.
  • Has a 10 mL capacity.
  • Constructed with clear material.
  • Designed with a flat bottom.
  • Supplied in quantities of 1000 per case.