Learn More
Agilent Technologies Agilent Technologies Vial Kit, 2 mL, Screw Top, Clear, Certified
Catalog No. NC3981667
Encompass
This certified vial kit contains 2 mL clear, screw top vials featuring a convenient write-on spot. Manufactured in an ISO 9001 certified environment, these vials are designed for reliable laboratory performance, with features such as reduced contamination packaging and compatibility with autosampler systems.
  • ISO 9001 certified manufacturing ensures high quality.
  • Packaged to reduce contamination.
  • Pierce tested with needles and syringes.
  • Automated vision system inspection for quality.
  • Compatible with autosampler gripping and injection.
  • Made from analytical grade 33 and 51 glass to prevent analyte removal.
  • Thicker glass walls prevent cracking.
  • Consistent lot-to-lot performance and rigorous quality control.
  • Integrated write-on spot for easy labeling.
  • Clear design for sample visibility.
  • 2 mL volume capacity.
  • Secure screw top.
Catalog No. NC3981667 Supplier Agilent Technologies Supplier No. 51906119
This certified vial kit contains 2 mL clear, screw top vials featuring a convenient write-on spot. Manufactured in an ISO 9001 certified environment, these vials are designed for reliable laboratory performance, with features such as reduced contamination packaging and compatibility with autosampler systems.
  • ISO 9001 certified manufacturing ensures high quality.
  • Packaged to reduce contamination.
  • Pierce tested with needles and syringes.
  • Automated vision system inspection for quality.
  • Compatible with autosampler gripping and injection.
  • Made from analytical grade 33 and 51 glass to prevent analyte removal.
  • Thicker glass walls prevent cracking.
  • Consistent lot-to-lot performance and rigorous quality control.
  • Integrated write-on spot for easy labeling.
  • Clear design for sample visibility.
  • 2 mL volume capacity.
  • Secure screw top.