Learn More
Agilent Technologies Vacuum Fluid Platinum, Pump Oil, For Rotary Vane Pumps, 946.353 mL
Catalog No. NC1799262
Encompass
This AVF Platinum vacuum pump fluid is a high-performance oil designed for use in rotary vane pumps for Agilent ICP-MS systems. It helps maintain required vacuum performance and ensures pumps meet specified operating standards.
  • Formulated for optimal performance in rotary vane pumps
  • Offers protection against corrosion
  • Provides excellent oxidative stability
  • Features low volatility for demanding analytical applications
  • Maintains a low pour point for reliable operation
  • Suitable for use in harsh chemical environments
  • Store at room temperature
Catalog No. NC1799262 Supplier Agilent Technologies Supplier No. X376064004
This AVF Platinum vacuum pump fluid is a high-performance oil designed for use in rotary vane pumps for Agilent ICP-MS systems. It helps maintain required vacuum performance and ensures pumps meet specified operating standards.
  • Formulated for optimal performance in rotary vane pumps
  • Offers protection against corrosion
  • Provides excellent oxidative stability
  • Features low volatility for demanding analytical applications
  • Maintains a low pour point for reliable operation
  • Suitable for use in harsh chemical environments
  • Store at room temperature