Learn More
Analytical Sales & Services AgileSLEEVE+ Flexible Capillary Heater – for 1/16″ OD Lines - 21
Catalog No. 5000805746
Encompass
AgileSLEEVE+ 21cm x 1/16" ID Capillary Heater
Catalog No. 50-008-05746 Supplier Analytical Sales & Services Supplier No. ASI2116H
AgileSLEEVE+ 21cm x 1/16" ID Capillary Heater