Learn More
Analytical Sales & Services AgileSLEEVE+ Flexible Capillary Heater – for 1/16″ OD Lines - 46
Catalog No. 5000806826
Encompass
AgileSLEEVE+ 46cm x 1/16" ID Capillary Heater
Catalog No. 50-008-06826 Supplier Analytical Sales & Services Supplier No. ASI4616H
AgileSLEEVE+ 46cm x 1/16" ID Capillary Heater