Learn More
Analytical Sales & Services AgileSLEEVE+ Flexible Capillary Heater – for 1/16″ OD Lines - 53
Catalog No. 5000807174
Encompass
AgileSLEEVE+ 53cm x 1/16" ID Capillary Heater
Catalog No. 50-008-07174 Supplier Analytical Sales & Services Supplier No. ASI5316H
AgileSLEEVE+ 53cm x 1/16" ID Capillary Heater