Learn More
Analytical Sales & Services AgileSLEEVE+ Flexible Capillary Heater – for 1/32″ OD Lines - 30
Catalog No. 5000806799
Encompass
AgileSLEEVE+ 30cm x 1/32" ID Capillary Heater
Catalog No. 50-008-06799 Supplier Analytical Sales & Services Supplier No. ASI3032H
AgileSLEEVE+ 30cm x 1/32" ID Capillary Heater