Learn More
AdipoGen Agistatin B
SDP
Supplier:  AdipoGen BVT0223M001
Encompass_Preferred
Chemical. CAS 144096-46-8. Formula C11H18O4. MW 214.3. Isolated from fungus A 6239. Tricyclic analog of agistatin A. Cholesterol biosynthesis inhibitor.
Catalog No. 50-256-2059