Learn More
AdipoGen Agistatin E
SDP
Catalog No. 502562053
Encompass_Preferred
Chemical. CAS 144096-48-0. Formula C11H16O5. MW 228.2. Isolated from fungus A 6239. Tricyclic analog of agistatin A . Cholesterol biosynthesis inhibitor. Mycotoxin.
Catalog No. 50-256-2053 Supplier AdipoGen Supplier No. BVT0231M001
Add to Cart
Add to Cart
Chemical. CAS 144096-48-0. Formula C11H16O5. MW 228.2. Isolated from fungus A 6239. Tricyclic analog of agistatin A . Cholesterol biosynthesis inhibitor. Mycotoxin.