Learn More
Chemglass Life Sciences Agitator Assembly For 15L Bio Reactors, 8mm OD Shaft, 390mm, M30 Thread
SDP
Catalog No. 501294478
Encompass
Agitator assembly is constructed of 316L stainless steel and screws into the M30 thread in the center of the headplate.Design ensures contamination-free sealing around the shaft.CLS-1385-05 attaches to the headplate with four bolts.CLS-1385-35 requires adapter plate, CLS-1385-30, for use in a 15L reactor.
Catalog No. 50-129-4478 Supplier Chemglass Life Sciences Supplier No. CLS138535
Agitator assembly is constructed of 316L stainless steel and screws into the M30 thread in the center of the headplate.Design ensures contamination-free sealing around the shaft.CLS-1385-05 attaches to the headplate with four bolts.CLS-1385-35 requires adapter plate, CLS-1385-30, for use in a 15L reactor.