Learn More
Chemglass Life Sciences Agitators, PTFE, High Viscosity, 25.4mm, 10" Blades, Solid One-Piece Design, Supplied with Safety Pin
SDP
Catalog No. NC1107168
Encompass_Preferred
Agitators, PTFE, High Viscosity, 25.4mm, 10" Blades, Solid One-Piece Design, Supplied with Safety Pin
Catalog No. NC1107168 Supplier Chemglass Life Sciences Supplier No. CG2096M12
Agitators, PTFE, High Viscosity, 25.4mm, 10" Blades, Solid One-Piece Design, Supplied with Safety Pin