Learn More
Chemglass Life Sciences Agitator, PTFE, High Viscosity, 5.5in Diameter, Fits 19mm Shaft Size, Fits 2L-5L Reactors
SDP
Catalog No. CG209608
Encompass_Preferred
Agitator, PTFE, High Viscosity, 5.5in Diameter, Fits 19mm Shaft Size, Fits 4L-5L Reactors. High viscosity agitator, made entirely of PTFE and glass filled PTFE, provides excellent stirring at the bottom of 4L through 5L reactors, especially when heavy slurries are being mixed. Agitator attaches to stir shaft by both a compression fitting and a glass filled PTFE safety pin. Heavy duty blades are 7mm thick. Supplied complete with one glass filled PTFE safety pin.
Catalog No. CG209608 Supplier Chemglass Life Sciences Supplier No. CG209608
Agitator, PTFE, High Viscosity, 5.5in Diameter, Fits 19mm Shaft Size, Fits 4L-5L Reactors. High viscosity agitator, made entirely of PTFE and glass filled PTFE, provides excellent stirring at the bottom of 4L through 5L reactors, especially when heavy slurries are being mixed. Agitator attaches to stir shaft by both a compression fitting and a glass filled PTFE safety pin. Heavy duty blades are 7mm thick. Supplied complete with one glass filled PTFE safety pin.