Learn More
Chemglass Life Sciences Agitator, PTFE, High Viscosity, 7.5in Diameter, Fits 19mm Shaft Size, Fits 10L-15L Reactors
SDP
Catalog No. 50872523
Encompass_Preferred
Agitator, PTFE, High Viscosity, 7.55in Diameter, Fits 19mm Shaft Size, Fits 10L-15L Reactors. High viscosity agitator, made entirely of PTFE and glass filled PTFE, provides excellent stirring at the bottom of 10L through 15L reactors, especially when heavy slurries are being mixed. Agitator attaches to stir shaft by both a compression fitting and a glass filled PTFE safety pin. Heavy duty blades are 7mm thick. Supplied complete with one glass filled PTFE safety pin.
Catalog No. 50-872-523 Supplier Chemglass Life Sciences Supplier No. CG209609
Agitator, PTFE, High Viscosity, 7.55in Diameter, Fits 19mm Shaft Size, Fits 10L-15L Reactors. High viscosity agitator, made entirely of PTFE and glass filled PTFE, provides excellent stirring at the bottom of 10L through 15L reactors, especially when heavy slurries are being mixed. Agitator attaches to stir shaft by both a compression fitting and a glass filled PTFE safety pin. Heavy duty blades are 7mm thick. Supplied complete with one glass filled PTFE safety pin.