Learn More
Ace Glass, Inc. Agitator, paddle, stainless steel, 10mm OD, 75mm diameter
SDP
Catalog No. NC0793116
Encompass
75mm diameter stainless steel multi-paddle agitator, for use with 10mm O.D. polished stainless steel or glass stirring shafts. One or more agitators can be positioned on the shaft at various locations, using supplied set screws to secure.
Catalog No. NC0793116 Supplier Ace Glass, Inc. Supplier No. 809423
75mm diameter stainless steel multi-paddle agitator, for use with 10mm O.D. polished stainless steel or glass stirring shafts. One or more agitators can be positioned on the shaft at various locations, using supplied set screws to secure.