Learn More
Ace Glass, Inc. PTFE agitator, (4) 50mm anchor style blades, fits 10mm knob style or drilled shafts
SDP
Catalog No. 5097645
Encompass_Preferred
Catalog No. 50-976-45 Supplier Ace Glass, Inc. Supplier No. 809102