Learn More
Ace Glass, Inc. PTFE agitator, (4) 152mm paddle style blades, fits 19mm knob style or drilled shafts. Replacement blades:(102mm)8091-14, (152mm) 8091-15
SDP
Catalog No. 50979671
Encompass_Preferred
Catalog No. 50-979-671 Supplier Ace Glass, Inc. Supplier No. 809120