Learn More
Chemglass Life Sciences Agitator, PTFE, 60°, 100mm OD Blade, Fits 16mm OD PTFE Covered SS Shafts
SDP
Catalog No. NC1730068
Encompass_Preferred
Agitator, PTFE, 60¿, 100mm OD Blade, Fits 16mm OD PTFE Covered SS Shafts. Agitator blades are constructed of PTFE with a glass filled PTFE locking nut. This enables them to be attached to the shaft in any vertical position. Note: Designed for use with CG-2079-X, CG-2078-X or CG-2096-X PTFE Covered stainless steel type shafts
Catalog No. NC1730068 Supplier Chemglass Life Sciences Supplier No. CG2095X164
Agitator, PTFE, 60¿, 100mm OD Blade, Fits 16mm OD PTFE Covered SS Shafts. Agitator blades are constructed of PTFE with a glass filled PTFE locking nut. This enables them to be attached to the shaft in any vertical position. Note: Designed for use with CG-2079-X, CG-2078-X or CG-2096-X PTFE Covered stainless steel type shafts