Learn More
Chemglass Life Sciences Agitator, PTFE, 60°, 50mm OD Blade, Fits 10mm OD PTFE Covered SS Shafts
SDP
Catalog No. 501531448
Encompass_Preferred
Agitator, PTFE, 60, 50mm OD Blade, Fits 10mm OD PTFE Covered SS Shafts. Agitator blades are constructed of PTFE with a glass filled PTFE locking nut. This enables them to be attached to the shaft in any vertical position. Note: Designed for use with CG-2079-X, CG-2078-X or CG-2096-X PTFE Covered Stainless Steel type shafts
Catalog No. 50-153-1448 Supplier Chemglass Life Sciences Supplier No. CG2095X102
Agitator, PTFE, 60, 50mm OD Blade, Fits 10mm OD PTFE Covered SS Shafts. Agitator blades are constructed of PTFE with a glass filled PTFE locking nut. This enables them to be attached to the shaft in any vertical position. Note: Designed for use with CG-2079-X, CG-2078-X or CG-2096-X PTFE Covered Stainless Steel type shafts