Learn More
Ace Glass, Inc. Agitator, turbine, PTFE, 10mm OD, 38mm diameter
SDP
Catalog No. 50976044
Encompass_Preferred
All-PTFE turbine type agitator for use with 10mm bead-type glass shafts or ACE 8071 PTFE shaft. Shaft opening is drilled straight thru to allow positioning of agitator at any drilled position on shaft as well as the bottom. Agitator has 10mm O.D. and 38mm diameter.
Catalog No. 50-976-044 Supplier Ace Glass, Inc. Supplier No. 809004
All-PTFE turbine type agitator for use with 10mm bead-type glass shafts or ACE 8071 PTFE shaft. Shaft opening is drilled straight thru to allow positioning of agitator at any drilled position on shaft as well as the bottom. Agitator has 10mm O.D. and 38mm diameter.