Learn More
Aviva Systems Biology Corp AGK Antibody - N-terminal region (ARP57169_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57169_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AGK. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human AGK. Alternate names: FLJ10842, MULK, MTDPS10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-271