Learn More
Cayman Chemical AGK2, 304896-28-4, 1 mg
SDP
Supplier:  Cayman Chemical 131451
Encompass_Preferred
A cell-permeable, selective inhibitor of SIRT2 (IC50 = 3.5 μM) that minimally affects either SIRT1 or SIRT3; rescues dopamine neurons from α-synuclein toxicity in both in vitro and in vivo Parkinson’s disease models
Catalog No. NC0700383