Learn More
Medchemexpress LLC HY-100578 5mg Medchemexpress, AGK2 CAS:304896-28-4 Purity:>98%
SDP
Catalog No. 501931890
Encompass_Preferred
Medchemexpress, HY-100578 5mg AGK2 CAS:304896-28-4 AGK2 is a selective SIRT2 inhibitor with IC 50 of 3.5 μM. AGK2 can also inhibit SIRT1 and SIRT3 with IC 50 of 30 and 91 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1890 Supplier Medchemexpress LLC Supplier No. HY1005785MG
Medchemexpress, HY-100578 5mg AGK2 CAS:304896-28-4 AGK2 is a selective SIRT2 inhibitor with IC 50 of 3.5 μM. AGK2 can also inhibit SIRT1 and SIRT3 with IC 50 of 30 and 91 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.