Learn More
Selleck Chemical LLC AGK2 5mg 304896-28-4
SDP
Supplier:  Selleck Chemical LLC S75775MG
Encompass_Preferred
AGK2 is a potent, and selective SIRT2 inhibitor with IC50 of 3.5 ?M that minimally affects either SIRT1 or SIRT3 at 10-fold higher levels. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4970