Learn More
Abcam AGK2, sirtuin2 (SIRT2) inhibitor, 10MG
Catalog No. 502706349
Encompass_Preferred
MW 434.3 Da, Purity >95%. Selective reversible cell-permeable sirtuin2 (SIRT2) inhibitor (IC₅₀ = 3.5 μM). Protects dopamine neurons from α-synuclein toxicity in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6349 Supplier Abcam Supplier No. AB14207310MG
MW 434.3 Da, Purity >95%. Selective reversible cell-permeable sirtuin2 (SIRT2) inhibitor (IC₅₀ = 3.5 μM). Protects dopamine neurons from α-synuclein toxicity in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement