Learn More
Enzo Life Sciences AGK7 (SIRT2 inhibitor, inactive control) (5mg). CAS: 304896-21-7
SDP
Catalog No. 502013448
Encompass_Preferred
Can be used as an inactive control in experiments with AGK2. Alternative name: SIRT2 Inhibitor (inactive control). Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3448 Supplier Enzo Life Sciences Supplier No. ENZCHM1460005
Can be used as an inactive control in experiments with AGK2. Alternative name: SIRT2 Inhibitor (inactive control). Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.