Learn More
Gold Biotechnology Inc AGL-1 Agrobacterium Electrocompetent Cells 5x50µl
SDP
Catalog No. NC2942865
Encompass_Preferred
GoldBio’s AGL-1 Agrobacterium Electrocompetent Cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction. Our AGL-1 strain harbors the C58 chromosomal backbone with an insertion mutation in its recA recombination gene. This mutation stabilizes recombinant plasmids. AGL-1 also has rifampicin and carbenicillin resistance genes in the genome useful for selection. A functional T-DNA binary system can be built using our AGL-1 strains as the T-DNA region has been deleted in the Ti plasmid pTiBO542 and instead it has a binary vector containing the missing T-region. The binary system makes possible to transfer genetic material into a host plant’s genome. Therefore, our system is often used for Agrobacterium-mediated transformation in mono and dicotyledonous species such as Arabidopsis thaliana, maize, and other plants.
Catalog No. NC2942865 Supplier Gold Biotechnology Inc Supplier No. CC2085X50C
GoldBio’s AGL-1 Agrobacterium Electrocompetent Cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction. Our AGL-1 strain harbors the C58 chromosomal backbone with an insertion mutation in its recA recombination gene. This mutation stabilizes recombinant plasmids. AGL-1 also has rifampicin and carbenicillin resistance genes in the genome useful for selection. A functional T-DNA binary system can be built using our AGL-1 strains as the T-DNA region has been deleted in the Ti plasmid pTiBO542 and instead it has a binary vector containing the missing T-region. The binary system makes possible to transfer genetic material into a host plant’s genome. Therefore, our system is often used for Agrobacterium-mediated transformation in mono and dicotyledonous species such as Arabidopsis thaliana, maize, and other plants.