Learn More
Gold Biotechnology Inc AGL 1 pSoup Agro Comp Cells
SDP
Catalog No. 5000562183
Encompass_Preferred
GoldBio s AGL-1 (pSoup) chemically competent Agrobacterium cells are optimized for high transformation efficiencies making them ideal for applications such as cDNA or gDNA library construction These cells are resistant to rifampicin carbenicillin and streptomycin providing versatility in selection and antibiotic resistance The Ti plasmid pTiBo542DT-DNA has been engineered to have the T-DNA region deleted which can be complemented with a binary vector containing the missing T-region to restore the T-DNA binary system function This feature enables efficient and precise transformation of plant genomes
Catalog No. 50-005-62183 Supplier Gold Biotechnology Inc Supplier No. CC11615X50
GoldBio s AGL-1 (pSoup) chemically competent Agrobacterium cells are optimized for high transformation efficiencies making them ideal for applications such as cDNA or gDNA library construction These cells are resistant to rifampicin carbenicillin and streptomycin providing versatility in selection and antibiotic resistance The Ti plasmid pTiBo542DT-DNA has been engineered to have the T-DNA region deleted which can be complemented with a binary vector containing the missing T-region to restore the T-DNA binary system function This feature enables efficient and precise transformation of plant genomes