Learn More
Gold Biotechnology Inc AGL 1 pSoup Agro Electro Cells
SDP
Catalog No. 5000562313
Encompass_Preferred
AGL-1 (pSoup) electrocompetent Agrobacterium cells are a highly efficient system for genetic transformation in various plant species This strain is derived from the C58 RecA-type Agrobacterium strain and carries the rifampicin carbenicillin and streptomycin resistance genes for selection purposes The Ti plasmid pTiBo542DT-DNA used in this system has a deleted T-DNA region and instead contains a binary vector that restores the T-DNA binary system function
Catalog No. 50-005-62313 Supplier Gold Biotechnology Inc Supplier No. CC21815X50
AGL-1 (pSoup) electrocompetent Agrobacterium cells are a highly efficient system for genetic transformation in various plant species This strain is derived from the C58 RecA-type Agrobacterium strain and carries the rifampicin carbenicillin and streptomycin resistance genes for selection purposes The Ti plasmid pTiBo542DT-DNA used in this system has a deleted T-DNA region and instead contains a binary vector that restores the T-DNA binary system function