Learn More
INTACT GENOMICS INC AGL1 (pSoup) Agrobacterium ElectroCompetent Cells 6x50µl
Catalog No. 5000082866
Encompass
Intact Genomics (ig) AGL-1 (pSoup) Agrobacterium ElectroCompetent Cells are optimized for the highest transformation efficiencies, ideal for applications with cDNA or gDNA library construction. The AGL1 (pSoup) strain has resistance to rifampicin, carbenicillin, and streptomycin (see genotype). The Ti plasmid pTiBo542DT-DNA has the T-DNA region deleted and transformed with a binary vector containing the missing T-region restores the T-DNA binary system function. The pSoup plasmid aids in the replication of pGreen, 62SK, and pGs2 series plasmids. This system is used for Agrobacterium-mediated transformation of dicots such as Arabidopsis thaliana, tobacco, potato, and monocots like corn.Product Includes and Storage:AGL-1 (pSoup) Agrobacterium ElectroComp Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCRecovery medium: 4 ºC
Catalog No. 50-000-82866 Supplier INTACT GENOMICS INC Supplier No. 1283PS12
Intact Genomics (ig) AGL-1 (pSoup) Agrobacterium ElectroCompetent Cells are optimized for the highest transformation efficiencies, ideal for applications with cDNA or gDNA library construction. The AGL1 (pSoup) strain has resistance to rifampicin, carbenicillin, and streptomycin (see genotype). The Ti plasmid pTiBo542DT-DNA has the T-DNA region deleted and transformed with a binary vector containing the missing T-region restores the T-DNA binary system function. The pSoup plasmid aids in the replication of pGreen, 62SK, and pGs2 series plasmids. This system is used for Agrobacterium-mediated transformation of dicots such as Arabidopsis thaliana, tobacco, potato, and monocots like corn.Product Includes and Storage:AGL-1 (pSoup) Agrobacterium ElectroComp Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCRecovery medium: 4 ºC