Learn More
Precision Glassblowing of Colorado AGLENT CHAMBER G1820-65337
SDP
Catalog No. 50420773
Encompass_Preferred
Axial Scott style Spray Chamber w/ Inner Tube flush with Outer Tube, Sample outlet is 90 deg. from drain, quartz
Catalog No. 50-420-773 Supplier Precision Glassblowing of Colorado Supplier No. PG90013
Axial Scott style Spray Chamber w/ Inner Tube flush with Outer Tube, Sample outlet is 90 deg. from drain, quartz