Learn More
Medchemexpress LLC HY-101238 100mg Medchemexpress, Agmatine (sulfate) CAS:2482-00-0 Purity:>98%
SDP
Catalog No. 501932072
Encompass_Preferred
Medchemexpress, HY-101238 100mg Agmatine (sulfate) CAS:2482-00-0 Agmatine sulfate exerts modulatory action at multiple molecular targets, such as neurotransmitter systems, ion channels and nitric oxide synthesis. It is an endogenous agonist at imidazoline receptor and a NO synthase inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2072 Supplier Medchemexpress LLC Supplier No. HY101238100MG
Medchemexpress, HY-101238 100mg Agmatine (sulfate) CAS:2482-00-0 Agmatine sulfate exerts modulatory action at multiple molecular targets, such as neurotransmitter systems, ion channels and nitric oxide synthesis. It is an endogenous agonist at imidazoline receptor and a NO synthase inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.