Learn More
Medchemexpress LLC Agmatine sulfate | 2482-00-0 | MFCD00013109 | 98.0% | 228.089 g/mol | C5H16N4O4S | 500 MG
SDP
Catalog No. 5000211525
Encompass_Preferred
Agmatine sulfate is an endogenous aminoguanidine derivative used in research to modulate imidazoline receptors, nitric oxide synthase, and related neurotransmitter systems. Supplied as a research-grade solid powder, the product is suitable for in vitro and preclinical studies and is reported at ~98.0% purity.
  • Research-grade solid powder suitable for in vitro studies.
  • Modulates imidazoline receptors and nitric oxide synthase.
  • Reported purity approximately 98.0%.
  • CAS number 2482-00-0 for unambiguous identification.
  • Commonly used in neurotransmitter and ion channel research applications.
Catalog No. 50-002-11525 Supplier Medchemexpress LLC Supplier No. HY101238500MG
Agmatine sulfate is an endogenous aminoguanidine derivative used in research to modulate imidazoline receptors, nitric oxide synthase, and related neurotransmitter systems. Supplied as a research-grade solid powder, the product is suitable for in vitro and preclinical studies and is reported at ~98.0% purity.
  • Research-grade solid powder suitable for in vitro studies.
  • Modulates imidazoline receptors and nitric oxide synthase.
  • Reported purity approximately 98.0%.
  • CAS number 2482-00-0 for unambiguous identification.
  • Commonly used in neurotransmitter and ion channel research applications.