Learn More
Medchemexpress LLC Agmatine sulfate | 2482-00-0 | MFCD00013109 | 98.0% | 228.27 g/mol | C5H16N4O4S | 1 G
SDP
Catalog No. 5000211748
Encompass_Preferred
Agmatine sulfate is a research-grade salt used in neurobiology and pharmacology to modulate imidazoline receptors and inhibit nitric oxide synthase. It is typically supplied as a white to off-white solid for in vitro studies, with recommended storage and handling instructions for preparing aqueous stocks.
  • High purity (98.0%).
  • White to off-white solid appearance.
  • Soluble in water ≥ 100 mg/mL; poorly soluble in DMSO.
  • Acts as an imidazoline receptor agonist and nitric oxide synthase inhibitor.
  • Suitable for in vitro pharmacology and receptor binding studies.
  • Store solid at 4°C; filter-sterilize aqueous stocks before use.
Catalog No. 50-002-11748 Supplier Medchemexpress LLC Supplier No. HY1012381G
Agmatine sulfate is a research-grade salt used in neurobiology and pharmacology to modulate imidazoline receptors and inhibit nitric oxide synthase. It is typically supplied as a white to off-white solid for in vitro studies, with recommended storage and handling instructions for preparing aqueous stocks.
  • High purity (98.0%).
  • White to off-white solid appearance.
  • Soluble in water ≥ 100 mg/mL; poorly soluble in DMSO.
  • Acts as an imidazoline receptor agonist and nitric oxide synthase inhibitor.
  • Suitable for in vitro pharmacology and receptor binding studies.
  • Store solid at 4°C; filter-sterilize aqueous stocks before use.