Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Agn 192403 hydrochloride | 1021868-90-5 | 99.0% | 189.73 g/mol | C10H20ClN | 10 MG
SDP

Catalog No. 5000212441
Encompass_Preferred

AGN 192403 hydrochloride is a research compound supplied as the hydrochloride salt and described as an I1-imidazoline receptor antagonist used in cardiovascular and neurological studies. It is provided as a solid with defined molecular properties and storage recommendations.

  • I1-imidazoline receptor antagonist for cardiovascular and neurological research.
  • CAS number 1021868-90-5.
  • Molecular formula C10H20ClN.
  • Molecular weight 189.73 g/mol.
  • Purity 99.0%.
  • Solid appearance; powder storage -20°C for long term, 4°C for short term.

Catalog No. 50-002-12441 Supplier Medchemexpress LLC Supplier No. HY101374A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AGN 192403 hydrochloride is a research compound supplied as the hydrochloride salt and described as an I1-imidazoline receptor antagonist used in cardiovascular and neurological studies. It is provided as a solid with defined molecular properties and storage recommendations.

  • I1-imidazoline receptor antagonist for cardiovascular and neurological research.
  • CAS number 1021868-90-5.
  • Molecular formula C10H20ClN.
  • Molecular weight 189.73 g/mol.
  • Purity 99.0%.
  • Solid appearance; powder storage -20°C for long term, 4°C for short term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.