Learn More
Medchemexpress LLC Agn 192403 hydrochloride | 175521-95-6 | >95.0% | 189.73 | C10H19N·HCl | 25 MG
SDP
Catalog No. 5000213208
Encompass_Preferred
AGN 192403 hydrochloride is a small-molecule I1 (imidazoline-1) receptor ligand used in cardiovascular and neurological research. It has been reported to show neuroprotective effects, and in cellular studies to bind the TMED9 cargo receptor, affecting MUC1-fs trafficking. The compound is used as a research tool to probe imidazoline receptor pharmacology and related mitochondrial pathways.
  • Selective I1 receptor activity suitable for receptor pharmacology studies.
  • Reported neuroprotective effects for neurological research applications.
  • Demonstrated interaction with TMED9, useful in cellular trafficking studies.
  • Applicable to in vitro assays and exploratory in vivo models.
  • Available from multiple research suppliers with supporting COA/technical data.
Catalog No. 50-002-13208 Supplier Medchemexpress LLC Supplier No. HY101374A25MG
AGN 192403 hydrochloride is a small-molecule I1 (imidazoline-1) receptor ligand used in cardiovascular and neurological research. It has been reported to show neuroprotective effects, and in cellular studies to bind the TMED9 cargo receptor, affecting MUC1-fs trafficking. The compound is used as a research tool to probe imidazoline receptor pharmacology and related mitochondrial pathways.
  • Selective I1 receptor activity suitable for receptor pharmacology studies.
  • Reported neuroprotective effects for neurological research applications.
  • Demonstrated interaction with TMED9, useful in cellular trafficking studies.
  • Applicable to in vitro assays and exploratory in vivo models.
  • Available from multiple research suppliers with supporting COA/technical data.