Learn More
Medchemexpress LLC HY-U00449 5mg Medchemexpress, AGN 193109 CAS:171746-21-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU004495MG
Encompass_Preferred
Medchemexpress, HY-U00449 5mg AGN 193109 CAS:171746-21-7 AGN 193109 is a retinoid analog, and acts as a specific and highly effective antagonist of retinoic acid receptors (RARs) , with K d s of 2 nM, 2 nM, and 3 nM for RARα , RARβ , and RARγ , respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4457