Learn More
Apexbio Technology LLC AGN 194310 229961-45-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064238
Encompass_Preferred
AGN 194310 is a pan-retinoic acid receptor (RAR) antagonist with affinity for RAR RAR and RAR It binds these receptors with Kd values of 3 nM 2 nM and 5 nM respectively By competing with RAR agonists AGN 194310 blocks receptor-mediated cellular responses In cell-based assays AGN 194310 inhibits the proliferation and colony formation of prostate carcinoma cell lines (LNCaP PC3 DU-145) induces G1 cell cycle arrest and promotes apoptosis In addition it suppresses agonist-induced differentiation in HL60 cells AGN 194310 serves as a research tool to investigate RAR-driven signaling pathways and tumor cell growth mechanisms particularly in oncology studies examining retinoid receptor functions
Catalog No. 50-000-64238 Supplier Apexbio Technology LLC Supplier No. B47025.1
 Restricted Item
Add to Cart
Add to Cart
AGN 194310 is a pan-retinoic acid receptor (RAR) antagonist with affinity for RAR RAR and RAR It binds these receptors with Kd values of 3 nM 2 nM and 5 nM respectively By competing with RAR agonists AGN 194310 blocks receptor-mediated cellular responses In cell-based assays AGN 194310 inhibits the proliferation and colony formation of prostate carcinoma cell lines (LNCaP PC3 DU-145) induces G1 cell cycle arrest and promotes apoptosis In addition it suppresses agonist-induced differentiation in HL60 cells AGN 194310 serves as a research tool to investigate RAR-driven signaling pathways and tumor cell growth mechanisms particularly in oncology studies examining retinoid receptor functions