Learn More
TARGETMOL CHEMICALS INC AGNUSIDE 10MG
SDP
Catalog No. 502580094
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Agnuside (chasteberry oil) inhibits COX-2 (IC50 0.026 mg/ml) but exhibits less than 10% inhibition of COX-1 at this concentration. It also inhibits 46.3 66.8 and 82.1% of P-glycoprotein (P-gp) ATPase activity at concentrations of 5 25 and 100 uM respectively. purity: 99%
Catalog No. 50-258-0094 Supplier TARGETMOL CHEMICALS INC Supplier No. T386810MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Agnuside (chasteberry oil) inhibits COX-2 (IC50 0.026 mg/ml) but exhibits less than 10% inhibition of COX-1 at this concentration. It also inhibits 46.3 66.8 and 82.1% of P-glycoprotein (P-gp) ATPase activity at concentrations of 5 25 and 100 uM respectively. purity: 99%