Learn More
Medchemexpress LLC HY-N2518 5mg Medchemexpress, Agnuside CAS:11027-63-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN25185MG
Encompass_Preferred
Medchemexpress, HY-N2518 5mg Agnuside CAS:11027-63-7 Agnuside is a compound isolated from Vitex negundo, down-regulates pro-inflammatory mediators PGE2 and LTB4, and reduces the expression of cytokines, with anti-arthritic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2901