Learn More
Medchemexpress LLC AGO1 Human sf9 Hi 50ug
SDP
Catalog No. 5000178679
Encompass_Preferred
The AGO1 protein is critical for RNA-mediated gene silencing (RNAi) by binding to short RNAs such as microRNAs (miRNAs) or short interfering RNAs (siRNAs) and inhibiting the translation of complementary mRNAs Despite lacking endonuclease activity AGO1 does not cleave target mRNA AGO1 Protein Human (sf9 His) is the recombinant human-derived AGO1 protein expressed by Sf9 insect cells with N-His labeled tag
Catalog No. 50-001-78679 Supplier Medchemexpress LLC Supplier No. HYP7442450UG
The AGO1 protein is critical for RNA-mediated gene silencing (RNAi) by binding to short RNAs such as microRNAs (miRNAs) or short interfering RNAs (siRNAs) and inhibiting the translation of complementary mRNAs Despite lacking endonuclease activity AGO1 does not cleave target mRNA AGO1 Protein Human (sf9 His) is the recombinant human-derived AGO1 protein expressed by Sf9 insect cells with N-His labeled tag