Learn More
Medchemexpress LLC Agomelatine | 138112-76-2 | >99.5% | 243.30 | 1 ML
SDP
Catalog No. 5000384498
Encompass_Preferred
Agomelatine, also known as S-20098, acts as a specific agonist for MT1 and MT2 receptors, demonstrating high affinity with Kis values ranging from 0.06 to 0.27 nM across various cell lines. It also functions as a selective antagonist for 5-HT2C receptors, exhibiting pKis of 6.4 and 6.2 at native and cloned human receptors, respectively. This product is intended for research purposes only.
  • Specific agonist of MT1 and MT2 receptors
  • Selective 5-HT2C receptor antagonist
  • High affinity for MT1 and MT2 receptors
  • Intended for research use only
Catalog No. 50-003-84498 Supplier Medchemexpress LLC Supplier No. HY17038B10MM/1ML
Agomelatine, also known as S-20098, acts as a specific agonist for MT1 and MT2 receptors, demonstrating high affinity with Kis values ranging from 0.06 to 0.27 nM across various cell lines. It also functions as a selective antagonist for 5-HT2C receptors, exhibiting pKis of 6.4 and 6.2 at native and cloned human receptors, respectively. This product is intended for research purposes only.
  • Specific agonist of MT1 and MT2 receptors
  • Selective 5-HT2C receptor antagonist
  • High affinity for MT1 and MT2 receptors
  • Intended for research use only