Learn More
Medchemexpress LLC Agomelatine-d3 | 1079389-38-0 | 98.0% | 246.32 | 1 MG
SDP
Catalog No. 5000384301
Encompass_Preferred
Agomelatine-d3 is the deuterium labeled version of Agomelatine. It functions as a specific agonist of MT1 and MT2 receptors and a selective 5-HT2C receptor antagonist.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-84301 Supplier Medchemexpress LLC Supplier No. HY17038S21MG
Agomelatine-d3 is the deuterium labeled version of Agomelatine. It functions as a specific agonist of MT1 and MT2 receptors and a selective 5-HT2C receptor antagonist.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.