Learn More
Medchemexpress LLC HY-17038A 5mg Medchemexpress, Agomelatine (hydrochloride) CAS:1176316-99-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17038A5MG
Encompass_Preferred
Medchemexpress, HY-17038A 5mg Agomelatine (hydrochloride) CAS:1176316-99-6 Agomelatine hydrochloride (S-20098 hydrochloride) is a specific agonist of MT1 and MT2 receptors with Kis of 0.1, 0.06, 0.12, and 0.27 nM for CHO-hMT1, HEK-hMT1, CHO-hMT2, and HEK-hMT2, respectively. Agomelatine hydrochloride is a selective 5-HT2C receptor antagonist with pKis of 6.4 and 6.2 at native (porcine) and cloned, human 5-HT2C receptors, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1932