Learn More
Cayman Chemical Agomelatn 10mg
SDP
Catalog No. 502953357
Encompass_Preferred
A melatonin receptor agonist (Kis = 0.14 and 0.41 nM for MT1 and MT2, respectively) and 5-HT2B and 5-HT2C antagonist (Kis = 0.26 and 0.71 nM, respectively); inhibits the 5-HT2-mediated penile erection response in rats at 40 mg/kg; increases extracellular levels of noradrenaline and dopamine in the frontal cortex of freely moving rats at 20-80 mg/kg; reduces immobility time in the forced swim test and increases the amount of time spent in the open arms of the elevated plus maze in mice in a transgenic neuroendocrine model of depression at 10 mg/kg
Catalog No. 50-295-3357 Supplier Cayman Chemical Supplier No. 1320310 MG
A melatonin receptor agonist (Kis = 0.14 and 0.41 nM for MT1 and MT2, respectively) and 5-HT2B and 5-HT2C antagonist (Kis = 0.26 and 0.71 nM, respectively); inhibits the 5-HT2-mediated penile erection response in rats at 40 mg/kg; increases extracellular levels of noradrenaline and dopamine in the frontal cortex of freely moving rats at 20-80 mg/kg; reduces immobility time in the forced swim test and increases the amount of time spent in the open arms of the elevated plus maze in mice in a transgenic neuroendocrine model of depression at 10 mg/kg