Learn More
Medchemexpress LLC Agouti-related Protein (AGRP) (83-132) Amide (human) TFA | 97.0% | 5676.57 (free base) | 1 MG
SDP
Catalog No. 5000400042
Encompass_Preferred
Agouti-related Protein (AGRP) (83-132) Amide (human) TFA is a fragment of agouti-related protein (AGRP) found in abundance in the arcuate nucleus of the hypothalamus. It primarily functions as an inverse agonist for the melanocortin-4 receptor (MC4R) to increase food intake.
  • Fragment of agouti-related protein (AGRP)
  • Acts as an inverse agonist for the melanocortin-4 receptor (MC4R)
  • Increases food intake and decreases spontaneous locomotor activity in rats
  • Solid, white to off-white appearance
  • Soluble in DMSO up to 100 mg/mL with ultrasonic assistance
  • Features a specific amino acid sequence with identified disulfide bridges
Catalog No. 50-004-00042 Supplier Medchemexpress LLC Supplier No. HYP3561A1MG
Agouti-related Protein (AGRP) (83-132) Amide (human) TFA is a fragment of agouti-related protein (AGRP) found in abundance in the arcuate nucleus of the hypothalamus. It primarily functions as an inverse agonist for the melanocortin-4 receptor (MC4R) to increase food intake.
  • Fragment of agouti-related protein (AGRP)
  • Acts as an inverse agonist for the melanocortin-4 receptor (MC4R)
  • Increases food intake and decreases spontaneous locomotor activity in rats
  • Solid, white to off-white appearance
  • Soluble in DMSO up to 100 mg/mL with ultrasonic assistance
  • Features a specific amino acid sequence with identified disulfide bridges