Learn More
Aviva Systems Biology Corp AGPAT2 Antibody - C-terminal region (ARP44637_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44637_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against AGPAT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human AGPAT2. Alternate names: 1-AGPAT2, BSCL, BSCL1, LPAAB, LPAAT-betaApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-53429